Assay Details
| Gene Symbol: MRPL33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003125284.5 | XP_003125332.1 | 149 | 65 | ||
| GenBank mRNA | AK344743.1 | - | 2 - 3 | 122 | 65 | |
| AY610506.1 | - | 2 - 3 | 70 | 65 | ||
More Information
Set Membership: |
|

MRPL33
Genomic Map