Assay Details
| Gene Symbol: GIMAP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013990746.2 | XP_013846200.2 | 158 | 63 | ||
| XM_021078913.1 | XP_020934572.1 | 170 | 63 | |||
| GenBank mRNA | AK345195.1 | - | 2 - 3 | 123 | 63 | |
More Information
Set Membership: |
|

GIMAP4
Genomic Map