Assay Details
| Gene Symbol: CCDC43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003131348.4 | XP_003131396.3 | 275 | 79 | ||
| XM_005668755.3 | XP_005668812.2 | 277 | 79 | |||
| GenBank mRNA | AK345337.1 | - | 1 - 2 | 218 | 79 | |
| AK395906.1 | - | 1 - 2 | 279 | 79 | ||
More Information
Set Membership: |
|

CCDC43
Genomic Map