Assay Details
| Gene Symbol: NGRN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021098987.1 | XP_020954646.1 | 2930 | 108 | ||
| GenBank mRNA | AK345808.1 | - | 2 - 3 | 302 | 108 | |
| AK346386.1 | - | 2 - 3 | 304 | 108 | ||
| AK350650.1 | - | 1 - 2 | 701 | 108 | ||
More Information
Set Membership: |
|

NGRN
Genomic Map