Assay Details
| Gene Symbol: LOC110259328 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021083598.1 | XP_020939257.1 | 86 | |||
| XM_021083599.1 | XP_020939258.1 | 86 | ||||
| XM_021083602.1 | XP_020939261.1 | 86 | ||||
| GenBank mRNA | AK346044.1 | - | 2 - 3 | 357 | 86 | |
| AK346052.1 | - | 2 - 3 | 357 | 86 | ||
| AK346397.1 | - | 2 - 3 | 86 | |||
| AK396606.1 | - | 2 - 3 | 86 | |||
More Information
Set Membership: |
|

LOC110259328
Genomic Map