Assay Details
| Gene Symbol: C6H16orf70 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021094040.1 | XP_020949699.1 | 88 | |||
| GenBank mRNA | AK346149.1 | - | 1 - 2 | 61 | 88 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021094040.1 transcripts.
Set Membership: |
|

C6H16orf70
Genomic Map