Assay Details
| Gene Symbol: HRC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003127287.5 | XP_003127335.2 | 1998 | 72 | ||
| GenBank mRNA | AK346278.1 | - | 4 - 5 | 1995 | 72 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: HRC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003127287.5 | XP_003127335.2 | 1998 | 72 | ||
| GenBank mRNA | AK346278.1 | - | 4 - 5 | 1995 | 72 | |
Set Membership: |
|