Assay Details
| Gene Symbol: C4BPB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003130455.6 | XP_003130503.1 | 928 | 65 | ||
| XM_013979643.2 | XP_013835097.1 | 1211 | 65 | |||
| GenBank mRNA | AK346434.1 | - | 5 - 6 | 1180 | 65 | |
More Information
Set Membership: |
|

C4BPB
Genomic Map