Assay Details
| Gene Symbol: VSTM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001924660.7 | XP_001924695.3 | 128 | 97 | ||
| XM_005671213.3 | XP_005671270.1 | 116 | 97 | |||
| XM_013983433.2 | XP_013838887.1 | 116 | 97 | |||
| XM_013983435.2 | XP_013838889.1 | 116 | 97 | |||
| GenBank mRNA | AK346451.1 | - | 1 - 2 | 91 | 97 | |
More Information
Set Membership: |
|

VSTM4
Genomic Map