Assay Details
| Gene Symbol: PDCL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003124899.6 | XP_003124947.3 | 740 | 65 | ||
| GenBank mRNA | AK347273.1 | - | 5 - 6 | 720 | 65 | |
| AK395482.1 | - | 5 - 6 | 718 | 65 | ||
| AY609578.1 | - | 5 - 6 | 615 | 65 | ||
More Information
Set Membership: |
|

PDCL3
Genomic Map