Assay Details
| Gene Symbol: AAGAB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003121743.5 | XP_003121791.5 | 611 | 69 | ||
| XM_005659864.3 | XP_005659921.3 | 611 | 69 | |||
| XM_005659865.3 | XP_005659922.3 | 650 | 69 | |||
| XM_005659866.3 | XP_005659923.3 | 611 | 69 | |||
| XM_021100396.1 | XP_020956055.1 | 752 | 69 | |||
| GenBank mRNA | AK347453.1 | - | 5 - 6 | 554 | 69 | |
| AK390002.1 | - | 5 - 6 | 586 | 69 | ||
| AK399554.1 | - | 5 - 6 | 542 | 69 | ||
| AK400722.1 | - | 5 - 6 | 554 | 69 | ||
More Information
Set Membership: |
|

AAGAB
Genomic Map