Assay Details
| Gene Symbol: P3H4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003131416.4 | XP_003131464.1 | 1600 | 75 | ||
| GenBank mRNA | AK347544.1 | - | 1 - 2 | 843 | 75 | |
| AK348342.1 | - | 2 - 3 | 1066 | 75 | ||
| AK350680.1 | - | 1 - 2 | 775 | 75 | ||
| AK396384.1 | - | 2 - 3 | 1073 | 75 | ||
| AY609530.1 | - | 2 - 3 | 1047 | 75 | ||
More Information
Set Membership: |
|

P3H4
Genomic Map