Assay Details
| Gene Symbol: TNFAIP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003483067.4 | XP_003483115.1 | 773 | 78 | ||
| GenBank mRNA | AK347774.1 | - | 4 - 5 | 676 | 78 | |
More Information
Set Membership: |
|

TNFAIP1
Genomic Map