Assay Details
| Gene Symbol: ZRANB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001929009.7 | XP_001929044.4 | 1961 | 81 | ||
| XM_005671552.3 | XP_005671609.1 | 673 | 81 | |||
| XM_013983854.2 | XP_013839308.1 | 1452 | 81 | |||
| XM_021073624.1 | XP_020929283.1 | 1961 | 81 | |||
| GenBank mRNA | AK348011.1 | - | 1 - 2 | 41 | 81 | |
More Information
Set Membership: |
|

ZRANB1
Genomic Map