Assay Details
| Gene Symbol: GNA15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005654729.3 | XP_005654786.3 | 4812 | 81 | ||
| GenBank mRNA | AK348023.1 | - | 3 - 4 | 897 | 81 | |
| AK350431.1 | - | 4 - 5 | 641 | 81 | ||
| AK395692.1 | - | 3 - 4 | 901 | 81 | ||
More Information
Set Membership: |
|

GNA15
Genomic Map