Assay Details
| Gene Symbol: RNF168 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021070157.1 | XP_020925816.1 | 594 | 67 | ||
| XM_021070158.1 | XP_020925817.1 | 581 | 67 | |||
| XM_021070159.1 | XP_020925818.1 | 606 | 67 | |||
| XM_021070160.1 | XP_020925819.1 | 829 | 67 | |||
| GenBank mRNA | AK348048.1 | - | 2 - 3 | 472 | 67 | |
| AK348777.1 | - | 2 - 3 | 429 | 67 | ||
| AK393718.1 | - | 2 - 3 | 556 | 67 | ||
More Information
Set Membership: |
|

RNF168
Genomic Map