Assay Details
| Gene Symbol: CCDC126 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003357478.4 | XP_003357526.1 | 617 | 108 | ||
| XM_021102513.1 | XP_020958172.1 | 797 | 108 | |||
| XM_021102514.1 | XP_020958173.1 | 645 | 108 | |||
| GenBank mRNA | AK348231.1 | - | 2 - 3 | 556 | 108 | |
More Information
Set Membership: |
|

CCDC126
Genomic Map