Assay Details
| Gene Symbol: SYP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003135078.5 | XP_003135126.2 | 186 | 63 | ||
| GenBank mRNA | AK349169.1 | - | 2 - 3 | 101 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: SYP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003135078.5 | XP_003135126.2 | 186 | 63 | ||
| GenBank mRNA | AK349169.1 | - | 2 - 3 | 101 | 63 | |
Set Membership: |
|