Assay Details
| Gene Symbol: C10H9orf64 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003357707.5 | XP_003357755.2 | 598 | 69 | ||
| XM_021064651.1 | XP_020920310.1 | 398 | 69 | |||
| XM_021064652.1 | XP_020920311.1 | 319 | 69 | |||
| GenBank mRNA | AK349179.1 | - | 1 - 2 | 587 | 69 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021064651.1, XM_021064652.1 transcripts.
Set Membership: |
|

C10H9orf64
Genomic Map