Assay Details
| Gene Symbol: C10H9orf64 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003357707.5 | XP_003357755.2 | 864 | 87 | ||
| XM_021064651.1 | XP_020920310.1 | 664 | 87 | |||
| XM_021064652.1 | XP_020920311.1 | 585 | 87 | |||
| GenBank mRNA | AK349179.1 | - | 2 - 3 | 853 | 87 | |
More Information
Set Membership: |
|

C10H9orf64
Genomic Map