Assay Details
| Gene Symbol: TUBB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK349184.1 | - | 2 - 3 | 217 | 110 | |
| AK391872.1 | - | 2 - 3 | 233 | 110 | ||
| AK400096.1 | - | 2 - 3 | 256 | 110 | ||
More Information
Set Membership: |
|

TUBB3
Genomic Map