Assay Details
| Gene Symbol: RAB39A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003129824.3 | XP_003129872.1 | 1258 | 86 | ||
| XM_021062738.1 | XP_020918397.1 | 1368 | 86 | |||
| GenBank mRNA | AK349449.1 | - | 1 - 2 | 383 | 86 | |
More Information
Set Membership: |
|

RAB39A
Genomic Map