Assay Details
| Gene Symbol: TMEM38A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021083580.1 | XP_020939239.1 | 768 | 75 | ||
| GenBank mRNA | AK349670.1 | - | 5 - 6 | 737 | 75 | |
More Information
Set Membership: |
|

TMEM38A
Genomic Map