Assay Details
| Gene Symbol: PSAPL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003482340.3 | XP_003482388.1 | 3906 | 104 | ||
| GenBank mRNA | AK349699.1 | - | 1 | 2121 | 104 | |
More Information
Set Membership: |
|

PSAPL1
Genomic Map