Assay Details
| Gene Symbol: TENT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003123752.6 | XP_003123800.1 | 1538 | 79 | ||
| XM_005661515.3 | XP_005661572.1 | 1486 | 79 | |||
| XM_021084486.1 | XP_020940145.1 | 1472 | 79 | |||
| XM_021084487.1 | XP_020940146.1 | 1460 | 79 | |||
| XM_021084488.1 | XP_020940147.1 | 1267 | 79 | |||
| XM_021084489.1 | XP_020940148.1 | 2640 | 79 | |||
| GenBank mRNA | AK350010.1 | - | 12 - 13 | 1452 | 79 | |
| AY609744.1 | - | 11 - 12 | 1181 | 79 | ||
More Information
Set Membership: |
|

TENT2
Genomic Map