Assay Details
| Gene Symbol: MANSC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK350031.1 | - | 1 - 2 | 77 | 59 | |
| AK396644.1 | - | 1 - 2 | 82 | 59 | ||
| AK397589.1 | - | 1 - 2 | 77 | 59 | ||
More Information
Set Membership: |
|

MANSC1
Genomic Map