Assay Details
| Gene Symbol: MANSC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021092342.1 | XP_020948001.1 | 398 | 77 | ||
| GenBank mRNA | AK350031.1 | - | 2 - 3 | 394 | 77 | |
| AK395063.1 | - | 2 - 3 | 77 | |||
| AK396644.1 | - | 2 - 3 | 77 | |||
| AK397589.1 | - | 2 - 3 | 394 | 77 | ||
More Information
Set Membership: |
|

MANSC1
Genomic Map