Assay Details
| Gene Symbol: SETD9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003133974.4 | XP_003134022.1 | 2017 | 81 | ||
| GenBank mRNA | AK350085.1 | - | 3 - 4 | 378 | 81 | |
More Information
Set Membership: |
|

SETD9
Genomic Map