Assay Details
| Gene Symbol: OTULIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003133841.4 | XP_003133889.1 | 1033 | 63 | ||
| XM_013984697.2 | XP_013840151.1 | 876 | 63 | |||
| GenBank mRNA | AK350162.1 | - | 6 - 7 | 999 | 63 | |
More Information
Set Membership: |
|

OTULIN
Genomic Map