Assay Details
| Gene Symbol: PARP10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003125470.6 | XP_003125518.4 | 3214 | 69 | ||
| GenBank mRNA | AK350599.1 | - | 8 - 9 | 2695 | 69 | |
More Information
Set Membership: |
|

PARP10
Genomic Map