Assay Details
| Gene Symbol: ZNF74 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021073672.1 | XP_020929331.1 | 3741 | 62 | ||
| GenBank mRNA | AK350687.1 | - | 1 | 1414 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: ZNF74 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021073672.1 | XP_020929331.1 | 3741 | 62 | ||
| GenBank mRNA | AK350687.1 | - | 1 | 1414 | 62 | |
Set Membership: |
|