Assay Details
| Gene Symbol: MCUR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003128183.4 | XP_003128231.1 | 540 | 50 | ||
| XM_005665577.3 | XP_005665634.1 | 540 | 50 | |||
| GenBank mRNA | AK350832.1 | - | 1 - 2 | 541 | 50 | |
| AK351046.1 | - | 1 - 2 | 541 | 50 | ||
More Information
Set Membership: |
|

MCUR1
Genomic Map