Assay Details
| Gene Symbol: CCDC162 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021090803.1 | XP_020946462.1 | 6198 | 103 | ||
| XM_021090805.1 | XP_020946464.1 | 6193 | 103 | |||
| XM_021090807.1 | XP_020946466.1 | 6548 | 103 | |||
| XM_021090809.1 | XP_020946468.1 | 6238 | 103 | |||
| XM_021090812.1 | XP_020946471.1 | 6588 | 103 | |||
| XM_021090816.1 | XP_020946475.1 | 6195 | 103 | |||
| XM_021090819.1 | XP_020946478.1 | 6189 | 103 | |||
| XM_021090827.1 | XP_020946486.1 | 6198 | 103 | |||
| XM_021090829.1 | XP_020946488.1 | 6195 | 103 | |||
| XM_021090832.1 | XP_020946491.1 | 5984 | 103 | |||
| XM_021090834.1 | XP_020946493.1 | 6060 | 103 | |||
| XM_021090836.1 | XP_020946495.1 | 5012 | 103 | |||
| GenBank mRNA | AK350846.1 | - | 7 - 8 | 856 | 103 | |
More Information
Set Membership: |
|

CCDC162
Genomic Map