Assay Details
| Gene Symbol: ANAPC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021081169.1 | XP_020936828.1 | 916 | 53 | ||
| GenBank mRNA | AK351281.1 | - | 912 | 53 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: ANAPC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021081169.1 | XP_020936828.1 | 916 | 53 | ||
| GenBank mRNA | AK351281.1 | - | 912 | 53 | ||
Set Membership: |
|