Assay Details
| Gene Symbol: GPR19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021092331.1 | XP_020947990.1 | 2973 | 97 | ||
| XM_021092332.1 | XP_020947991.1 | 2660 | 97 | |||
| XM_021092333.1 | XP_020947992.1 | 1023 | 97 | |||
| XM_021092335.1 | XP_020947994.1 | 785 | 97 | |||
| GenBank mRNA | AK351879.1 | - | 3 - 4 | 347 | 97 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021092331.1, XM_021092332.1, XM_021092333.1, XM_021092335.1 transcripts.
Set Membership: |
|

GPR19
Genomic Map