Assay Details
| Gene Symbol: SSBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021078594.1 | XP_020934253.1 | 369 | 96 | ||
| XM_021078595.1 | XP_020934254.1 | 374 | 96 | |||
| XM_021078596.1 | XP_020934255.1 | 379 | 96 | |||
| XM_021078597.1 | XP_020934256.1 | 356 | 96 | |||
| XM_021078598.1 | XP_020934257.1 | 353 | 96 | |||
| XM_021078599.1 | XP_020934258.1 | 374 | 96 | |||
| XM_021078600.1 | XP_020934259.1 | 438 | 96 | |||
| XM_021078601.1 | XP_020934260.1 | 374 | 96 | |||
| GenBank mRNA | AK352341.1 | - | 3 - 4 | 438 | 96 | |
| AK391310.1 | - | 3 - 4 | 379 | 96 | ||
More Information
Set Membership: |
|

SSBP1
Genomic Map