Assay Details
| Gene Symbol: FBXO44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005664993.2 | XP_005665050.1 | 499 | 88 | ||
| XM_021095338.1 | XP_020950997.1 | 590 | 88 | |||
| GenBank mRNA | AK389325.1 | - | 1 - 2 | 111 | 88 | |
More Information
Set Membership: |
|

FBXO44
Genomic Map