Assay Details
| Gene Symbol: TTC9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005656367.3 | XP_005656424.2 | 1151 | 76 | ||
| GenBank mRNA | AK389398.1 | - | 3 - 4 | 588 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: TTC9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005656367.3 | XP_005656424.2 | 1151 | 76 | ||
| GenBank mRNA | AK389398.1 | - | 3 - 4 | 588 | 76 | |
Set Membership: |
|