Assay Details
| Gene Symbol: TXNL4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001924851.4 | XP_001924886.1 | 373 | 99 | ||
| XM_013993974.2 | XP_013849428.1 | 422 | 99 | |||
| GenBank mRNA | AK231597.1 | - | 3 - 4 | 341 | 99 | |
More Information
Set Membership: |
|

TXNL4A
Genomic Map