Assay Details
| Gene Symbol: WDR54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003124992.4 | XP_003125040.1 | 478 | 62 | ||
| XM_005662440.1 | XP_005662497.1 | 468 | 62 | |||
| GenBank mRNA | AK231910.1 | - | 4 - 5 | 449 | 62 | |
| AK236347.1 | - | 4 - 5 | 410 | 62 | ||
| AK398256.1 | - | 4 - 5 | 425 | 62 | ||
More Information
Set Membership: |
|

WDR54
Genomic Map