Assay Details
| Gene Symbol: BIN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003483383.4 | XP_003483431.1 | 411 | 65 | ||
| XM_021073387.1 | XP_020929046.1 | 652 | 65 | |||
| XM_021073388.1 | XP_020929047.1 | 650 | 65 | |||
| XM_021073389.1 | XP_020929048.1 | 412 | 65 | |||
| XM_021073390.1 | XP_020929049.1 | 643 | 65 | |||
| GenBank mRNA | AK231954.1 | - | 5 - 6 | 371 | 65 | |
| AK398576.1 | - | 5 - 6 | 372 | 65 | ||
More Information
Set Membership: |
|

BIN3
Genomic Map