Assay Details
| Gene Symbol: DNASE1L3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021069068.1 | XP_020924727.1 | 1200 | 61 | ||
| GenBank mRNA | AK231967.1 | - | 7 - 8 | 968 | 61 | |
| AK346544.1 | - | 7 - 8 | 887 | 61 | ||
| AK394448.1 | - | 7 - 8 | 969 | 61 | ||
More Information
Set Membership: |
|

DNASE1L3
Genomic Map