Assay Details
| Gene Symbol: SIT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005660221.3 | XP_005660278.1 | 265 | 61 | ||
| GenBank mRNA | AK232017.1 | - | 1 - 2 | 219 | 61 | |
| AY609768.1 | - | 195 | 61 | |||
More Information
Set Membership: |
|

SIT1
Genomic Map