Assay Details
| Gene Symbol: TTLL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003481524.3 | XP_003481572.1 | 1069 | 58 | ||
| XM_003481525.4 | XP_003481573.1 | 1009 | 58 | |||
| XM_021091413.1 | XP_020947072.1 | 726 | 58 | |||
| XM_021091414.1 | XP_020947073.1 | 768 | 58 | |||
| XM_021091415.1 | XP_020947074.1 | 1058 | 58 | |||
| XM_021091416.1 | XP_020947075.1 | 962 | 58 | |||
| XM_021091417.1 | XP_020947076.1 | 936 | 58 | |||
| XM_021091418.1 | XP_020947077.1 | 1059 | 58 | |||
| GenBank mRNA | AK232042.1 | - | 8 - 9 | 993 | 58 | |
| AK236231.1 | - | 9 - 10 | 1076 | 58 | ||
| AK395378.1 | - | 9 - 10 | 1053 | 58 | ||
More Information
Set Membership: |
|

TTLL1
Genomic Map