Assay Details
| Gene Symbol: PPTC7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003132884.4 | XP_003132932.2 | 3889 | 110 | ||
| GenBank mRNA | AK389675.1 | - | 1 | 1625 | 110 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: PPTC7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003132884.4 | XP_003132932.2 | 3889 | 110 | ||
| GenBank mRNA | AK389675.1 | - | 1 | 1625 | 110 | |
Set Membership: |
|