Assay Details
| Gene Symbol: RAB39B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003135517.5 | XP_003135565.1 | 3842 | 108 | ||
| GenBank mRNA | AK389850.1 | - | 1 | 1318 | 108 | |
More Information
Set Membership: |
|

RAB39B
Genomic Map