Assay Details
| Gene Symbol: ANKH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003359727.5 | XP_003359775.2 | 962 | 67 | ||
| GenBank mRNA | AK389951.1 | - | 1 - 2 | 46 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: ANKH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003359727.5 | XP_003359775.2 | 962 | 67 | ||
| GenBank mRNA | AK389951.1 | - | 1 - 2 | 46 | 67 | |
Set Membership: |
|