Assay Details
| Gene Symbol: GABRA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003134089.6 | XP_003134137.4 | 1633 | 73 | ||
| GenBank mRNA | AK390387.1 | - | 8 - 9 | 1179 | 73 | |
More Information
Set Membership: |
|

GABRA1
Genomic Map