Assay Details
| Gene Symbol: CAVIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021066983.1 | XP_020922642.1 | 6334 | 65 | ||
| GenBank mRNA | AK391022.1 | - | 1 | 221 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: CAVIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021066983.1 | XP_020922642.1 | 6334 | 65 | ||
| GenBank mRNA | AK391022.1 | - | 1 | 221 | 65 | |
Set Membership: |
|