Assay Details
| Gene Symbol: LIX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003123811.5 | XP_003123859.3 | 6635 | 84 | ||
| GenBank mRNA | AK391083.1 | - | 4 - 5 | 600 | 84 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: LIX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003123811.5 | XP_003123859.3 | 6635 | 84 | ||
| GenBank mRNA | AK391083.1 | - | 4 - 5 | 600 | 84 | |
Set Membership: |
|